Ethyl 4-oxo-4H-pyran-2-carboxylate 97%

Supplier: Ambeed
Warning

A176897-100MG A176897-250MG
76813-402EA 51.99 USD
76813-402 76813-404
Ethyl 4-oxo-4H-pyran-2-carboxylate 97%
Ethyl 4-oxo-4H-pyran-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD17012130
CAS Number: 1551-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR