(3R,4R)-rel-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride 98%

Supplier: Ambeed
Warning

A176842-100G A176842-1G A176842-25G A176842-5G
76813-394EA 494.38 USD
76813-394 76813-396 76813-398 76813-400
(3R,4R)-rel-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride 98%
(3R,4R)-rel-1-Benzyl-N,4-dimethylpiperidin-3-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂·2HCl
MW: 291.26 g/mol
MDL Number: MFCD11044501
CAS Number: 477600-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR