2-Hydrazinylpyridine hydrochloride 97%

Supplier: Ambeed
Warning

A176517-250MG A176517-1G
76813-384EA 6 USD
76813-384 77671-506
2-Hydrazinylpyridine hydrochloride 97%
2-Hydrazinylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈ClN₃
MDL Number: MFCD20275393
CAS Number: 51169-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR