4-(1-Methyl-1H-pyrazol-5-yl)pyridine 98%

Supplier: Ambeed
Warning

A176489-100MG A176489-1G A176489-250MG
76813-378EA 19 USD
76813-378 76813-380 76813-382
4-(1-Methyl-1H-pyrazol-5-yl)pyridine 98%
4-(1-Methyl-1H-pyrazol-5-yl)pyridine

Precaución: for Research Use Only

Formula: C₉H₉N₃
MDL Number: MFCD20275476
CAS Number: 905281-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR