4-Bromo-N-phenylbenzenesulphonamide 98%

Supplier: Ambeed
Warning

A176364-1G A176364-250MG A176364-5G
76813-372EA 38 USD
76813-372 76813-374 76813-376
4-Bromo-N-phenylbenzenesulphonamide 98%
4-Bromo-N-phenylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₂S
MDL Number: MFCD00159375
CAS Number: 7454-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR