5-Phenylmorpholin-3-one 95%

Supplier: Ambeed
Warning

A174835-100MG A174835-1G A174835-250MG A174835-5G
76813-326EA 14 USD
76813-326 76813-328 76813-330 76813-332
5-Phenylmorpholin-3-one 95%
5-Phenylmorpholin-3-one
Formula: C₁₀H₁₁NO₂
MDL Number: MFCD18250283
CAS Number: 1260672-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR