5-Iodo-2-methylpyridin-4-amine 95%

Supplier: Ambeed
Warning

A173674-100MG A173674-1G A173674-250MG
76813-298EA 91.24 USD
76813-298 76813-300 76813-302
5-Iodo-2-methylpyridin-4-amine 95%
5-Iodo-2-methylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₆H₇IN₂
MW: 234.03 g/mol
MDL Number: MFCD11040268
CAS Number: 849353-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR