tert-Butyl 3-(methylamino)piperidine-1-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A173547-100MG A173547-250MG
76813-294EA 12.74 USD
76813-294 76813-296
tert-Butyl 3-(methylamino)piperidine-1-carboxylate hydrochloride 95%
tert-Butyl 3-(methylamino)piperidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₃ClN₂O₂
MW: 250.77 g/mol
MDL Number: MFCD09878919
CAS Number: 1187927-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR