5,7-Dichlorothieno[3,2-b]pyridine 98%

Supplier: Ambeed
Warning

A172468-100MG A172468-250MG
76813-250EA 44.56 USD
76813-250 76813-252
5,7-Dichlorothieno[3,2-b]pyridine 98%
5,7-Dichlorothieno[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NS
MW: 204.0764 g/mol
MDL Number: MFCD09953620
CAS Number: 74695-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR