4-Fluoro-1-indanone 95%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2,3-dihydro-1H-inden-1-one

A172296-100MG A172296-1G A172296-250MG A172296-25G A172296-5G
76813-240EA 6 USD
76813-240 76813-242 76813-244 76813-246 76813-248
4-Fluoro-1-indanone 95%
4-Fluoro-1-indanone

Precaución: for Research Use Only

Formula: C₉H₇FO
MDL Number: MFCD00797829
CAS Number: 699-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR