Methyl 6-chloro-4-methylpyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloro-4-methylpicolinate

A171425-100MG A171425-1G A171425-250MG A171425-5G A171425-25G
76813-220EA 8.49 USD
76813-220 76813-222 76813-224 76813-226 77671-440
Methyl 6-chloro-4-methylpyridine-2-carboxylate 98%
Methyl 6-chloro-4-methylpyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
MDL Number: MFCD14582002
CAS Number: 1186605-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR