4’-Bromo-3’-methylacetophenone 98%

Supplier: Ambeed
Warning

A171263-10G A171263-1G A171263-250MG A171263-5G A171263-25G
76813-212EA 132 USD
76813-212 76813-214 76813-216 77671-432 77883-321
4’-Bromo-3’-methylacetophenone 98%
4’-Bromo-3’-methylacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD00045017
CAS Number: 37074-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR