5-Bromo-2-methoxyquinoline-8-carbaldehyde 95%

Supplier: Ambeed
Warning

A170458-100MG A170458-10G A170458-1G A170458-250MG A170458-5G
76813-188EA 85.94 USD
76813-188 76813-190 76813-192 76813-194 76813-196
5-Bromo-2-methoxyquinoline-8-carbaldehyde 95%
5-Bromo-2-methoxyquinoline-8-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO₂
MW: 266.09 g/mol
MDL Number: MFCD21603865
CAS Number: 885687-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR