Ethyl (R)-(-)-2-pyrrolidone-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Pyr-OEt

A170025-100G A170025-1G A170025-25G A170025-5G A170025-10G
76813-176EA 95.49 USD
76813-176 76813-178 76813-180 76813-182 77671-410
Ethyl (R)-(-)-2-pyrrolidone-5-carboxylate 97%
Ethyl (R)-(-)-2-pyrrolidone-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.167 g/mol
MDL Number: MFCD00010848
CAS Number: 68766-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR