(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride 98%

Supplier: Ambeed
Warning

A169765-1MG A169765-1G A169765-250MG
76813-172EA 5 USD
76813-172 77883-310 77883-311
(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride 98%
(S)-1-(2,4-Difluorophenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClF₂N
MDL Number: MFCD11101244
CAS Number: 844647-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR