2-Bromo-3-nitroaniline 95%

Supplier: Ambeed
Warning

A169418-250MG A169418-100MG A169418-5G
76813-168EA 103 USD
76813-168 77883-307 77883-308
2-Bromo-3-nitroaniline 95%
2-Bromo-3-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MDL Number: MFCD12923157
CAS Number: 35757-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR