2,4-Dibromo-6-methylpyridine 95%

Supplier: Ambeed
Warning

A169285-100MG A169285-1G A169285-5G A169285-250MG
76813-158EA 27.59 USD
76813-158 76813-160 76813-162 77671-400
2,4-Dibromo-6-methylpyridine 95%
2,4-Dibromo-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
MDL Number: MFCD09909845
CAS Number: 79055-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR