N(α)-Cbz-D-histidine 97%

Supplier: Ambeed
Warning

Synonyms: Z-D-His-OH, N(α)-Benzyloxycarbonyl-D-histidine

A168905-100MG A168905-250MG
76813-136EA 7.43 USD
76813-136 76813-138
N(α)-Cbz-D-histidine 97%
N(α)-Cbz-D-histidine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N₃O₄
MW: 289.3 g/mol
MDL Number: MFCD00067702
CAS Number: 67424-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR