Ethyl-3-cyclopropyl-1-methyl-1H-pyrazole-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-cyclopropyl-1-methyl-1H-pyrazole-5-carboxylate

A168034-100MG A168034-1G A168034-250MG
76813-100EA 55.17 USD
76813-100 76813-102 76813-104
Ethyl-3-cyclopropyl-1-methyl-1H-pyrazole-5-carboxylate 95%
Ethyl-3-cyclopropyl-1-methyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₂
MW: 194.24 g/mol
MDL Number: MFCD15072123
CAS Number: 133261-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR