4-Methoxy-2,3-dihydro-1H-isoindol-1-one 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxyisoindolin-1-one

A167996-100MG A167996-10G A167996-1G A167996-250MG A167996-5G
76813-090EA 14.86 USD
76813-090 76813-092 76813-094 76813-096 76813-098
4-Methoxy-2,3-dihydro-1H-isoindol-1-one 95%
4-Methoxy-2,3-dihydro-1H-isoindol-1-one

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.173 g/mol
MDL Number: MFCD09701287
CAS Number: 366453-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR