tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate 97%

Supplier: Ambeed
Warning

Synonyms: 3-[2-[2-(2-Hydroxyethoxy)ethoxy]ethoxy]propionic acid tert-Butyl ester, Hydroxy-PEG3-(CH2)2-Boc, 12-Hydroxy-4,7,10-trioxadodecanoic acid tert-Butyl ester, tert-Butyl 3-[2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy]propionate

A167809-10G A167809-1G A167809-25G A167809-5G A167809-100G A167809-250MG
76813-082EA 71 USD
76813-082 76813-084 76813-086 76813-088 77671-376 77883-302
tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate 97%
tert-Butyl 12-Hydroxy-4,7,10-trioxadodecanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₆O₆
MW: 278.35 g/mol
MDL Number: MFCD03791106
CAS Number: 186020-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR