2-Pyridyl tribromomethyl sulfone 98%

Supplier: Ambeed
Warning

Synonyms: 2-((Tribromomethyl)sulphonyl)pyridine, 2-(Tribromomethanesulphonyl)pyridine

A167615-1G A167615-5G
76813-076EA 14.86 USD
76813-076 76813-078
2-Pyridyl tribromomethyl sulfone 98%
2-Pyridyl tribromomethyl sulfone

Precaución: for Research Use Only

Formula: C₆H₄Br₃NO₂S
MW: 393.88 g/mol
MDL Number: MFCD02093493
CAS Number: 59626-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR