3,5-Diamino-6-chloropyrazine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A167446-1G A167446-250MG A167446-5G
76813-060EA 51.99 USD
76813-060 76813-062 76813-064
3,5-Diamino-6-chloropyrazine-2-carboxylic acid 98%
3,5-Diamino-6-chloropyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅ClN₄O₂
MW: 188.572 g/mol
MDL Number: MFCD06208349
CAS Number: 4878-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR