5'-Deoxy-5-fluorocytidine 97%
Supplier: Ambeed
|
Warning
|
A166600-10G
A166600-1G
A166600-250MG
A166600-5G
A166600-25G
A166600-100G
76813-046EA
23
USD
76813-046
76813-048
76813-050
76813-052
77611-622
77671-352
5'-Deoxy-5-fluorocytidine 97%
5'-Deoxy-5-fluorocytidine
Precaución: for Research Use Only
Formula:
C₉H₁₂FN₃O₄ |
MDL Number:
MFCD07369278 CAS Number: 66335-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...