6-Bromopyrazolo[1,5-a]pyridine 97%

Supplier: Ambeed
Warning

A166578-100MG A166578-10G A166578-1G A166578-250MG A166578-5G
76813-032EA 15.92 USD
76813-032 76813-034 76813-036 76813-038 76813-040
6-Bromopyrazolo[1,5-a]pyridine 97%
6-Bromopyrazolo[1,5-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.032 g/mol
MDL Number: MFCD18633056
CAS Number: 1264193-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR