PHA-680632 98%

Supplier: Ambeed
Warning

Synonyms: N-(2,6-diethylphenyl)-3-[[4-(4-methylpiperazin-1-yl)benzoyl]amino]-4,6-dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxamide, N-(2,6-Diethylphenyl)-3-(4-(4-methylpiperazin-1-yl)benzamido)-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxamide

A165443-10MG A165443-1MG A165443-25MG A165443-50MG A165443-5MG
76812-978EA 58.35 USD
76812-978 76812-980 76812-982 76812-984 76812-986
PHA-680632 98%
PHA-680632

Precaución: for Research Use Only

Formula: C₂₈H₃₅N₇O₂
MW: 501.62 g/mol
MDL Number: MFCD16038900
CAS Number: 398493-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR