2-((2-(2-Hydroxyethoxy)-4-nitrophenyl)amino)ethanol 98%

Supplier: Ambeed
Warning

A165309-10G A165309-5G
76812-968EA 25.47 USD
76812-968 76812-970
2-((2-(2-Hydroxyethoxy)-4-nitrophenyl)amino)ethanol 98%
2-((2-(2-Hydroxyethoxy)-4-nitrophenyl)amino)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₅
MDL Number: MFCD03265655
CAS Number: 59820-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR