3-Iodo-1H-pyrrolo[2,3-c]pyridine 98%

Supplier: Ambeed
Warning

A165282-1G A165282-250MG
76812-964EA 44.56 USD
76812-964 76812-966
3-Iodo-1H-pyrrolo[2,3-c]pyridine 98%
3-Iodo-1H-pyrrolo[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MDL Number: MFCD10699183
CAS Number: 956003-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR