4-Chloro-3-fluoro-2-methylpyridine 95%

Supplier: Ambeed
Warning

A164933-1G A164933-250MG
76812-960EA 151 USD
76812-960 76812-962
4-Chloro-3-fluoro-2-methylpyridine 95%
4-Chloro-3-fluoro-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MDL Number: MFCD16610395
CAS Number: 1195251-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR