3,6-Difluoro-2-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A164899-1G A164899-250MG A164899-5G
76812-954EA 46.68 USD
76812-954 76812-956 76812-958
3,6-Difluoro-2-methoxybenzaldehyde 98%
3,6-Difluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD08275289
CAS Number: 887267-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR