1-Methyl-1H-pyrazol-5(4H)-one 97%

Supplier: Ambeed
Warning

A164251-10G A164251-1G A164251-250MG A164251-25G A164251-5G
76812-930EA 103.97 USD
76812-930 76812-932 76812-934 76812-936 76812-938
1-Methyl-1H-pyrazol-5(4H)-one 97%
1-Methyl-1H-pyrazol-5(4H)-one

Precaución: for Research Use Only

Formula: C₄H₆N₂O
MW: 98.10 g/mol
MDL Number: MFCD20229439
CAS Number: 10234-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR