tert-Butyl-2-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate

A164118-100MG A164118-1G A164118-250MG A164118-25G A164118-5G
76812-918EA 40.32 USD
76812-918 76812-920 76812-922 76812-924 76812-926
tert-Butyl-2-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate 95%
tert-Butyl-2-chloro-7,8-dihydro-1,6-naphthyridine-6(5H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₇ClN₂O₂
MW: 268.75 g/mol
MDL Number: MFCD10565958
CAS Number: 1151665-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR