Methyl-2-(3-fluorophenyl)acetate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(3-fluorophenyl)acetate, Methyl 3-Fluorophenylacetate, 3-Fluorophenylacetic acid methyll ester, Methyl 3-fluorophenylacetate

A164115-100G A164115-10G A164115-1G A164115-25G A164115-5G
76812-908EA 78 USD
76812-908 76812-910 76812-912 76812-914 76812-916
Methyl-2-(3-fluorophenyl)acetate 97%
Methyl-2-(3-fluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD03093288
CAS Number: 64123-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR