trans-3-Hydroxycyclobutanecarboxylic acid 97%

Supplier: Ambeed
Warning

A163664-100MG A163664-250MG
76812-886EA 59.42 USD
76812-886 76812-888
trans-3-Hydroxycyclobutanecarboxylic acid 97%
trans-3-Hydroxycyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₈O₃
MW: 116.115 g/mol
MDL Number: MFCD17676395
CAS Number: 1268521-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR