[3,3'-Bipyridin]-6-amine 97%

Supplier: Ambeed
Warning

A163631-1G A163631-250MG A163631-100MG
76812-882EA 160 USD
76812-882 76812-884 77611-592
[3,3'-Bipyridin]-6-amine 97%
[3,3'-Bipyridin]-6-amine

Precaución: for Research Use Only

Formula: C₁₀H₉N₃
MDL Number: MFCD12033341
CAS Number: 31970-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR