4-Bromo-N,N-dimethylaniline 98%

Supplier: Ambeed
Warning

A163174-100G A163174-10G A163174-25G A163174-500G A163174-1000G
76812-856EA 32 USD
76812-856 76812-858 77320-040 77611-590 77883-274
4-Bromo-N,N-dimethylaniline 98%
4-Bromo-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MDL Number: MFCD00000093
CAS Number: 586-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR