Methyl (3R,5S,E)-7-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)-3,5-dihydroxyhept-6-enoate 95%

Supplier: Ambeed
Warning

A162812-1G A162812-250MG A162812-5G
76812-844EA 18.04 USD
76812-844 76812-846 76812-848
Methyl (3R,5S,E)-7-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)-3,5-dihydroxyhept-6-enoate 95%
Methyl (3R,5S,E)-7-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)-3,5-dihydroxyhept-6-enoate

Precaución: for Research Use Only

Formula: C₂₃H₃₀FN₃O₆S
MW: 495.56 g/mol
MDL Number: MFCD12756002
CAS Number: 147118-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR