Dicyclohexyl(2'-methoxy-[1,1'-biphenyl]-2-yl)phosphine 97%

Supplier: Ambeed
Warning

A162066-1G A162066-250MG A162066-25G A162066-5G
76812-800EA 10 USD
76812-800 76812-802 76812-804 76812-806
Dicyclohexyl(2'-methoxy-[1,1'-biphenyl]-2-yl)phosphine 97%
Dicyclohexyl(2'-methoxy-[1,1'-biphenyl]-2-yl)phosphine

Precaución: for Research Use Only

Formula: C₂₅H₃₃OP
MW: 380.50 g/mol
MDL Number: MFCD19441234
CAS Number: 255835-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR