Methyl 2-chloro-3-nitrobenzoate 97%

Supplier: Ambeed
Warning

A161787-100MG A161787-250MG A161787-5G A161787-1G
76812-790EA 13.8 USD
76812-790 76812-792 77567-362 77671-294
Methyl 2-chloro-3-nitrobenzoate 97%
Methyl 2-chloro-3-nitrobenzoate

Precaución: For research use only.

Formula: C₈H₆ClNO₄
MW: 215.591 g/mol
MDL Number: MFCD00482383
CAS Number: 53553-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR