2-Amino-5-iodonicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-iodopyridine-3-carboxylic acid

A161630-1G A161630-250MG A161630-5G
76812-784EA 20.16 USD
76812-784 76812-786 76812-788
2-Amino-5-iodonicotinic acid 97%
2-Amino-5-iodonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅IN₂O₂
MW: 264.02 g/mol
MDL Number: MFCD10568356
CAS Number: 54400-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR