LY-310762 hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-(4-(4-Fluorobenzoyl)piperidin-1-yl)ethyl)-3,3-dimethylindolin-2-one hydrochloride, LY310762, 1-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-1,3-dihydro-3,3-dimethyl-2H-indol-2-one hydrochloride

A161442-100MG A161442-10MG A161442-1MG A161442-250MG A161442-50MG A161442-5MG
76812-762EA 149.59 USD
76812-762 76812-764 76812-766 76812-768 76812-770 76812-772
LY-310762 hydrochloride 98%
LY-310762 hydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₂₇FN₂O₂.HCl
MW: 430.9 g/mol
MDL Number: MFCD07772272
CAS Number: 192927-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR