4-Chloro-2-phenylpyrimidine 95%

Supplier: Ambeed
Warning

A161267-250MG A161267-1G A161267-100MG
76812-754EA 33 USD
76812-754 77671-278 77881-261
4-Chloro-2-phenylpyrimidine 95%
4-Chloro-2-phenylpyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂
MDL Number: MFCD00234601
CAS Number: 14790-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR