2,6-Difluorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluorobenzeneboronic acid pinacol ester, 2,6-Difluorophenylboronic acid, pinacol ester, 2-(2,6-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A161032-100G A161032-10G A161032-1G A161032-250MG A161032-25G A161032-5G
76812-740EA 366.02 USD
76812-740 76812-742 76812-744 76812-746 76812-748 76812-750
2,6-Difluorophenylboronic acid pinacol ester 98%
2,6-Difluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₂O₂
MW: 240.06 g/mol
MDL Number: MFCD09801043
CAS Number: 863868-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR