2,6-Dimethyl-1H-indole 97%

Supplier: Ambeed
Warning

A160860-1G A160860-250MG A160860-100MG
76812-730EA 103 USD
76812-730 76812-732 77671-276
2,6-Dimethyl-1H-indole 97%
2,6-Dimethyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₀H₁₁N
MDL Number: MFCD04117975
CAS Number: 5649-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR