(S)-Methyl pyrrolidine-3-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A160566-100MG A160566-1G A160566-250MG A160566-25G A160566-5G
76812-696EA 31.83 USD
76812-696 76812-698 76812-700 76812-702 76812-704
(S)-Methyl pyrrolidine-3-carboxylate hydrochloride 98%
(S)-Methyl pyrrolidine-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂·ClH
MW: 165.618 g/mol
MDL Number: MFCD11840196
CAS Number: 1099646-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR