6-Chloro-5-methylnicotinonitrile 98%

Supplier: Ambeed
Warning

A160279-10G A160279-1G A160279-250MG A160279-25G A160279-5G
76812-684EA 74 USD
76812-684 76812-686 76812-688 76812-690 76812-692
6-Chloro-5-methylnicotinonitrile 98%
6-Chloro-5-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MDL Number: MFCD09261082
CAS Number: 66909-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR