5-Chloro-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile 95%

Supplier: Ambeed
Warning

A159870-1G A159870-250MG
76812-664EA 65.78 USD
76812-664 76812-666
5-Chloro-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile 95%
5-Chloro-1-(pyridin-2-yl)-1H-pyrazole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅ClN₄
MW: 204.62 g/mol
MDL Number: MFCD17168530
CAS Number: 104771-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR