5-Bromo-2-thiophenesulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromothiophene-2-sulphonamide

A159565-1G A159565-25G A159565-5G
76812-652EA 6.37 USD
76812-652 76812-654 76812-656
5-Bromo-2-thiophenesulfonamide 97%
5-Bromo-2-thiophenesulfonamide

Precaución: for Research Use Only

Formula: C₄H₄BrNO₂S₂
MW: 242.11 g/mol
MDL Number: MFCD00067990
CAS Number: 53595-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR