7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline 98%
Supplier: Ambeed
|
Warning
|
A159272-100G
A159272-10G
A159272-1G
A159272-250MG
A159272-25G
A159272-5G
76812-630EA
220
USD
76812-630
76812-632
76812-634
76812-636
76812-638
76812-640
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline 98%
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO |
MDL Number:
MFCD00627122 CAS Number: 1810-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...