Methyl 5-bromo-2-oxoindoline-7-carboxylate 95%

Supplier: Ambeed
Warning

A159194-100MG A159194-250MG A159194-5G
76812-626EA 39 USD
76812-626 76812-628 77567-310
Methyl 5-bromo-2-oxoindoline-7-carboxylate 95%
Methyl 5-bromo-2-oxoindoline-7-carboxylate

Precaución: For research use only.

Formula: C₁₀H₈BrNO₃
MDL Number: MFCD08458993
CAS Number: 898747-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR